Merck
  • Chronic activation of hexosamine biosynthesis in the heart triggers pathological cardiac remodeling.
Chronic activation of hexosamine biosynthesis in the heart triggers pathological cardiac remodeling.
Nature communications (2020-04-15)
Diem Hong Tran, Herman I May, Qinfeng Li, Xiang Luo, Jian Huang, Guangyu Zhang, Erica Niewold, Xiaoding Wang, Thomas G Gillette, Yingfeng Deng, Zhao V Wang
ABSTRACT
The hexosamine biosynthetic pathway (HBP) plays critical roles in nutrient sensing, stress response, and cell growth. However, its contribution to cardiac hypertrophic growth and heart failure remains incompletely understood. Here, we show that the HBP is induced in cardiomyocytes during hypertrophic growth. Overexpression of Gfat1 (glutamine:fructose-6-phosphate amidotransferase 1), the rate-limiting enzyme of HBP, promotes cardiomyocyte growth. On the other hand, Gfat1 inhibition significantly blunts phenylephrine-induced hypertrophic growth in cultured cardiomyocytes. Moreover, cardiac-specific overexpression of Gfat1 exacerbates pressure overload-induced cardiac hypertrophy, fibrosis, and cardiac dysfunction. Conversely, deletion of Gfat1 in cardiomyocytes attenuates pathological cardiac remodeling in response to pressure overload. Mechanistically, persistent upregulation of the HBP triggers decompensated hypertrophy through activation of mTOR while Gfat1 deficiency shows cardioprotection and a concomitant decrease in mTOR activity. Taken together, our results reveal that chronic upregulation of the HBP under hemodynamic stress induces pathological cardiac hypertrophy and heart failure through persistent activation of mTOR.
Sigma-Aldrich
Monoclonal Anti-PGM3 antibody produced in mouse, clone 1E2-1B12, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-DSCR1 (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, powder
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-GNPNAT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution