Merck
  • Icariin increases chondrocyte vitality by promoting hypoxia-inducible factor-1α expression and anaerobic glycolysis.
Icariin increases chondrocyte vitality by promoting hypoxia-inducible factor-1α expression and anaerobic glycolysis.
The Knee (2019-12-31)
Pengzhen Wang, Xifeng Xiong, Jinli Zhang, Shengnan Qin, Wen Wang, Zhihe Liu
ABSTRACT
Articular cartilage is a unique avascular tissue in which chondrocytes are embedded in extracellular matrix (ECM). The decreased ECM resulting from the loss of articular chondrocyte viability leads to degenerative diseases such as osteoarthritis (OA). This study aims to investigate the effect of icariin (ICA) on ECM synthesis and chondrocyte viability. Micromass culture, alcian blue, and Safran O (SO)/fast green staining were used to investigate chondrocyte viability and ECM synthesis in chondrocytes treated with ICA. The expression of hypoxia-inducible factor-1α (HIF-1α), SOX9, and anaerobic glycolysis enzymes were detected by western blot and reverse transcription-quantitative polymerase chain reaction. ICA, an active flavonoid component of Herba epimedii, was demonstrated to increase chondrocyte viability and ECM synthesis. HIF-1α is a key mediator of chondrocyte response to fluctuations in oxygen availability during cartilage development or damage, and its expression was unregulated by ICA treatment. Meanwhile, ICA treatment increased SOX9 expression, which is a key regulator of ECM synthesis. Furthermore, ICA treatment increased the expression of glucose transporter 1 (GLUT1), glucose-6-phosphate dehydrogenase (G6PD), phosphoglycerate kinase 1 (PGK1), and pyruvate dehydrogenase kinase 1 (PDK1), which contribute to glucose transfer and anaerobic glycolysis. The present study revealed that ICA treatment facilitates chondrocyte vitality by promoting HIF-1α expression and anaerobic glycolysis. Therefore, ICA could be a novel clinical treatment for OA.
Sigma-Aldrich
Nunc® EasYFlasks, Nunclon® Δ Surface, Nunclon® Δ surface treatment, 75 cm2 angled neck, filter cap, 100/cs
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Xylazine, ≥99%