Merck
  • T cell receptor and cytokine signal integration in CD8+ T cells is mediated by the protein Themis.
T cell receptor and cytokine signal integration in CD8+ T cells is mediated by the protein Themis.
Nature immunology (2020-01-15)
Joanna Brzostek, Namrata Gautam, Xiang Zhao, Elijah W Chen, Monika Mehta, Desmond W H Tung, Yen Leong Chua, Jiawei Yap, Su H Cho, Shvetha Sankaran, Vasily Rybakin, Guo Fu, Nicholas R J Gascoigne
ABSTRACT
T cell homeostasis and functional responsiveness require signals from self-peptide-major histocompatibility complex (self-pMHC) and cytokines, but the mechanisms controlling this signal integration are unknown. Using a conditional deletion of the T cell lineage-specific protein Themis, we show that Themis is required for the maintenance of peripheral CD8+ T cells and for proliferative CD8+ T cell responses to low-affinity pMHC aided by cytokines. Themis-deficient peripheral T cells show a phenotype indicative of reduced tonic signaling from self-pMHC, strongly suggesting that Themis is a positive regulator of T cell receptor signal strength in response to low-affinity self-pMHC in peripheral T cells. Signals from low-affinity pMHC and cytokines synergistically induce phosphorylation of the kinase Akt, metabolic changes and c-Myc transcription factor induction in CD8+ T cells only in the presence of Themis. This function of Themis is mediated through Shp1 phosphatase, as peripheral Themis and Shp1 double deletion rescues the peripheral CD8+ T cell maintenance.
Product Description
Millipore
Immobilon®-FL PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane with low background fluorescence for Western blotting. Compatible with visible and infrared fluorescent probes.
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)