Merck
  • Tissue engineering of retina through high resolution 3-dimensional inkjet bioprinting.
Tissue engineering of retina through high resolution 3-dimensional inkjet bioprinting.
Biofabrication (2019-10-03)
Elahe Masaeli, Valérie Forster, Serge Picaud, Fereshte Karamali, Mohammad Hossein Nasr-Esfahani, Christophe Marquette
ABSTRACT
The mammalian retina contains multiple cellular layers, each carrying out a specific task. Such a controlled organization should be considered as a crucial factor for designing retinal therapies. The maintenance of retinal layered complexity through the use of scaffold-free techniques has recently emerged as a promising approach for clinical ocular tissue engineering. In an attempt to fabricate such layered retinal model, we are proposing herein a unique inkjet bioprinting system applied to the deposition of a photoreceptor cells (PRs) layer on top of a bioprinted retinal pigment epithelium (RPE), in a precise arrangement and without any carrier material. The results showed that, after bioprinting, both RPE and PRs were well positioned in a layered structure and expressed their structural markers, which was further demonstrated by ZO1, MITF, rhodopsin, opsin B, opsin R/G and PNA immunostaining, three days after bioprinting. We also showed that considerable amounts of human vascular endothelial growth factors (hVEGF) were released from the RPE printed layer, which confirmed the formation of a functional RPE monolayer after bioprinting. Microstructures of bioprinted cells as well as phagocytosis of photoreceptor outer segments by apical RPE microvilli were finally established through transmission electron microscopy (TEM) imaging. In summary, using this carrier-free bioprinting method, it was possible to develop a reasonable in vitro retina model for studying some sight-threatening diseases, such as age-related macular degeneration (AMD) and retinitis pigmentosa (RP).
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D.E.R. 332, used as embedding medium
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Gelatin methacryloyl, gel strength 300 g Bloom, 80% degree of substitution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Resazurin based
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TissueFab® Discrete GelMA, 90-110 bloom, 50% degree of substitution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TissueFab® Discrete GelMA, 300 bloom, 80% degree of substitution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TissueFab® Discrete GelMA, 170-195 bloom, 50% degree of substitution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TissueFab® Discrete GelMA, 300 bloom, 50% degree of substitution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, laboratory grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, for molecular biology