Merck
  • Interleukin-1 beta-modulated gene expression in immortalized human chondrocytes.
Interleukin-1 beta-modulated gene expression in immortalized human chondrocytes.
The Journal of clinical investigation (1994-12-01)
M B Goldring, J R Birkhead, L F Suen, R Yamin, S Mizuno, J Glowacki, J L Arbiser, J F Apperley
ABSTRACT
Immortalized human chondrocytes were established by transfection of primary cultures of juvenile costal chondrocytes with vectors encoding simian virus 40 large T antigen and selection in suspension culture over agarose. Stable cell lines were generated that exhibited chondrocyte morphology, continuous proliferative capacity (> 80 passages) in monolayer culture in serum-containing medium, and expression of mRNAs encoding chondrocyte-specific collagens II, IX, and XI and proteoglycans in an insulin-containing serum substitute. They did not express type X collagen or versican mRNA. These cells synthesized and secreted extracellular matrix molecules that were reactive with monoclonal antibodies against type II collagen, large proteoglycan (PG-H, aggrecan), and chondroitin-4- and chondroitin-6-sulfate. Interleukin-1 beta (IL-1 beta) decreased the levels of type II collagen mRNA and increased the levels of mRNAs for collagenase, stromelysin, and immediate early genes (egr-1, c-fos, c-jun, and jun-B). These cell lines also expressed reporter gene constructs containing regulatory sequences (-577/+3,428 bp) of the type II collagen gene (COL2A1) in transient transfection experiments, and IL-1 beta suppressed this expression by 50-80%. These results show that immortalized human chondrocytes displaying cartilage-specific modulation by IL-1 beta can be used as a model for studying normal and pathological repair mechanisms.
Sigma-Aldrich
C20A4 Human Chondrocyte Cell Line, C20A4 Human Chondrocyte Cell Line is widely used as a model cell line for studying normal and pathological cartilage repair mechanisms related to chondrocyte biology and physiology.
Sigma-Aldrich
C28/I2 Human Chondrocyte Cell Line, C28/I2 Human Chondrocyte Cell Line is widely used as a model cell line for studying normal and pathological cartilage repair mechanisms related to chondrocyte biology and physiology.
Sigma-Aldrich
TC28a2 Human Chondrocyte Cell Line, TC28a2 Human Chondrocyte Cell Line is widely used as a model cell line for studying normal and pathological cartilage repair mechanisms related to chondrocyte biology and physiology.
Sigma-Aldrich
Anti-Collagen Type II Antibody, clone COLL-II, ascites fluid, clone COLL-II, Chemicon®