Merck
  • Discovery of Mcl-1-specific inhibitor AZD5991 and preclinical activity in multiple myeloma and acute myeloid leukemia.
Discovery of Mcl-1-specific inhibitor AZD5991 and preclinical activity in multiple myeloma and acute myeloid leukemia.
Nature communications (2018-12-19)
Adriana E Tron, Matthew A Belmonte, Ammar Adam, Brian M Aquila, Lawrence H Boise, Elisabetta Chiarparin, Justin Cidado, Kevin J Embrey, Eric Gangl, Francis D Gibbons, Gareth P Gregory, David Hargreaves, J Adam Hendricks, Jeffrey W Johannes, Ricky W Johnstone, Steven L Kazmirski, Jason G Kettle, Michelle L Lamb, Shannon M Matulis, Ajay K Nooka, Martin J Packer, Bo Peng, Philip B Rawlins, Daniel W Robbins, Alwin G Schuller, Nancy Su, Wenzhan Yang, Qing Ye, Xiaolan Zheng, J Paul Secrist, Edwin A Clark, David M Wilson, Stephen E Fawell, Alexander W Hird
ABSTRACT
Mcl-1 is a member of the Bcl-2 family of proteins that promotes cell survival by preventing induction of apoptosis in many cancers. High expression of Mcl-1 causes tumorigenesis and resistance to anticancer therapies highlighting the potential of Mcl-1 inhibitors as anticancer drugs. Here, we describe AZD5991, a rationally designed macrocyclic molecule with high selectivity and affinity for Mcl-1 currently in clinical development. Our studies demonstrate that AZD5991 binds directly to Mcl-1 and induces rapid apoptosis in cancer cells, most notably myeloma and acute myeloid leukemia, by activating the Bak-dependent mitochondrial apoptotic pathway. AZD5991 shows potent antitumor activity in vivo with complete tumor regression in several models of multiple myeloma and acute myeloid leukemia after a single tolerated dose as monotherapy or in combination with bortezomib or venetoclax. Based on these promising data, a Phase I clinical trial has been launched for evaluation of AZD5991 in patients with hematological malignancies (NCT03218683).
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Anti-Mcl-1 Antibody, clone RC13, clone RC13, from mouse
Pricing and availability is not currently available. Contact Technical Service