Merck
  • Comprehensive in vivo identification of the c-Myc mRNA protein interactome using HyPR-MS.
Comprehensive in vivo identification of the c-Myc mRNA protein interactome using HyPR-MS.
RNA (New York, N.Y.) (2019-07-13)
Michele Spiniello, Maisie I Steinbrink, Anthony J Cesnik, Rachel M Miller, Mark Scalf, Michael R Shortreed, Lloyd M Smith
ABSTRACT
Proteins bind mRNA through their entire life cycle from transcription to degradation. We analyzed c-Myc mRNA protein interactors in vivo using the HyPR-MS method to capture the crosslinked mRNA by hybridization and then analyzed the bound proteins using mass spectrometry proteomics. Using HyPR-MS, 229 c-Myc mRNA-binding proteins were identified, confirming previously proposed interactors, suggesting new interactors, and providing information related to the roles and pathways known to involve c-Myc. We performed structural and functional analysis of these proteins and validated our findings with a combination of RIP-qPCR experiments, in vitro results released in past studies, publicly available RIP- and eCLIP-seq data, and results from software tools for predicting RNA-protein interactions.
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
EZ-Magna NuCLEAR RIP (Cross-Linked) Nuclear RNA-Binding Protein Immunoprecipitation Kit, EZ-Magna Nuclear RIP (Cross-Linked) RNA-Binding Protein Immunoprecipitation Kit is designed for the analysis of chromatin associated RNA such lncRNAs, enhancer RNAs and miRNAs.