Merck
  • Silencing of KCNA1 suppresses the cervical cancer development via mitochondria damage.
Silencing of KCNA1 suppresses the cervical cancer development via mitochondria damage.
Channels (Austin, Tex.) (2019-07-30)
Li Liu, Yumei Chen, Qingyuan Zhang, Changzhong Li
ABSTRACT
Voltage-gated potassium channel subfamily A member 1 (KCNA1/Kv1.1) is an important component of type A potassium channels, which has been found to be involved in various tumors. This study aimed to identify the role of KCNA1 in cervical cancer and explore the related mechanism. The levels of KCNA1 in cervical cancer tissues and cell lines were examined by Western blot and qPCR. Cell proliferation and invasion were assessed by CCK-8 and transwell assays, respectively. Protein levels of Hedgehog (Hhg), Wnt and Notch were detected by Western blot. The mitochondrial capacity was examined by immunostaining with MitoTracker Red CMXRos. KCNA1 was highly expressed in cervical cancer tissues and cell lines, and correlated with poor prognosis. In addition, depletion of KCNA1 suppressed growth, proliferation, migration and invasion of HeLa cells. Moreover, KCNA1 could regulate the Hhg, Wnt and Notch signaling pathways and cause mitochondrial dysfunction. The present study has demonstrated that KCNA1 is an oncogene excessively expressed in cervical cancer, and promotes tumor progression by regulating the Hhg, Wnt and Notch signaling pathways and the mitochondrial capacity. Therefore, our results provide a theoretical basis for the discovery of novel clinical treatment against cervical cancer.
Sigma-Aldrich
Monoclonal Anti-NOTCH3 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-WNT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-KCNA1 antibody produced in rabbit, affinity isolated antibody