Merck
  • The Inhibition of miR-144-3p on Cell Proliferation and Metastasis by Targeting TOP2A in HCMV-Positive Glioblastoma Cells.
The Inhibition of miR-144-3p on Cell Proliferation and Metastasis by Targeting TOP2A in HCMV-Positive Glioblastoma Cells.
Molecules (Basel, Switzerland) (2018-12-14)
Jingyi Song, Qingxia Ma, Ming Hu, Dongmeng Qian, Bin Wang, Ningning He
ABSTRACT
Glioblastoma (GBM), the most common type of primary tumor in the central nervous system, is a very aggressive brain tumor with poor prognosis and a high recurrence rate. Increasing evidence suggests that human cytomegalovirus (HCMV) infection is related to GBM and leads to GBM cell growth and metastasis. MicroRNAs are important regulators in the growth and metastasis of glioblastoma. This study aimed to demonstrate the role of miR-144-3p in HCMV-positive glioblastoma. We found that, after HCMV infection, the expression of miR-144-3p decreased, whereas the expression of TOP2A increased. Bioinformatics analyses indicated that miR-144-3p directly targets the TOP2A 3'-UTR (Untranslated Region). We discovered that the overexpression of miR-144-3p downregulated the overexpression of TOP2A and inhibited the proliferation, clone formation, and invasion of HCMV-positive glioma in vitro. Taken together, these results show that miR-144-3p inhibited growth and promoted apoptosis in glioma cells by targeting TOP2A.
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
EZ-Magna NuCLEAR RIP (Cross-Linked) Nuclear RNA-Binding Protein Immunoprecipitation Kit, EZ-Magna Nuclear RIP (Cross-Linked) RNA-Binding Protein Immunoprecipitation Kit is designed for the analysis of chromatin associated RNA such lncRNAs, enhancer RNAs and miRNAs.