Merck
  • Stable reduction of STARD4 alters cholesterol regulation and lipid homeostasis.
Stable reduction of STARD4 alters cholesterol regulation and lipid homeostasis.
Biochimica et biophysica acta. Molecular and cell biology of lipids (2020-01-10)
David B Iaea, Zachary R Spahr, Rajesh K Singh, Robin B Chan, Bowen Zhou, Rohan Bareja, Olivier Elemento, Gilbert Di Paolo, Xiaoxue Zhang, Frederick R Maxfield
ABSTRACT
STARD4, a member of the evolutionarily conserved START gene family, is a soluble sterol transport protein implicated in cholesterol sensing and maintenance of cellular homeostasis. STARD4 is widely expressed and has been shown to transfer sterol between liposomes as well as organelles in cells. However, STARD4 knockout mice lack an obvious phenotype, so the overall role of STARD4 is unclear. To model long term depletion of STARD4 in cells, we use short hairpin RNA technology to stably decrease STARD4 expression in human U2OS osteosarcoma cells (STARD4-KD). We show that STARD4-KD cells display increased total cholesterol, slower cholesterol trafficking between the plasma membrane and the endocytic recycling compartment, and increased plasma membrane fluidity. These effects can all be rescued by transient expression of a short hairpin RNA-resistant STARD4 construct. Some of the cholesterol increase was due to excess storage in late endosomes or lysosomes. To understand the effects of reduced STARD4, we carried out transcriptional and lipidomic profiling of control and STARD4-KD cells. Reduction of STARD4 activates compensatory mechanisms that alter membrane composition and lipid homeostasis. Based on these observations, we propose that STARD4 functions as a critical sterol transport protein involved in sterol sensing and maintaining lipid homeostasis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Avanti
12:0 SM (d18:1/12:0), Avanti Research - A Croda Brand 860583P, powder
Avanti
13:0 Lyso PI, 1-tridecanoyl-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt), powder
Avanti
16:0-18:0-16:0 D5 TG, Avanti Research - A Croda Brand 860902P, powder
Avanti
cholesterol-d7, Avanti Research - A Croda Brand
Avanti
14:0 PG, Avanti Research - A Croda Brand
Avanti
C17 Ceramide (d18:1/17:0), Avanti Research - A Croda Brand
Avanti
C12 Glucosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860543P, powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Avanti
14:0 Lyso PE, 1-myristoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine, powder
Avanti
C12 Galactosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860544P, powder
Avanti
C12 Lactosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860545P, powder
Avanti
17:0 Lyso PC, 1-heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
17:0 Lyso PC, Avanti Research - A Croda Brand
Avanti
4ME 16:0 Diether DG, Avanti Research - A Croda Brand 999986C
Avanti
4ME 16:0 Diether DG, Avanti Research - A Croda Brand 999986O