Merck
  • Subunit composition, crystallization and preliminary crystallographic studies of the Desulfovibrio gigas aldehyde oxidoreductase containing molybdenum and [2Fe-2S] centers.
Subunit composition, crystallization and preliminary crystallographic studies of the Desulfovibrio gigas aldehyde oxidoreductase containing molybdenum and [2Fe-2S] centers.
European journal of biochemistry (1993-08-01)
M J Romão, B A Barata, M Archer, K Lobeck, I Moura, M A Carrondo, J LeGall, F Lottspeich, R Huber, J J Moura
ABSTRACT
The Desulfovibrio gigas aldehyde oxidoreductase contains molybdenum bound to a pterin cofactor and [2Fe-2S] centers. The enzyme was characterized by SDS/PAGE, gel-filtration and analytical ultracentrifugation experiments. It was crystallized at 4 degrees C, pH 7.2, using isopropanol and MgCl2 as precipitants. The crystals diffract beyond 0.3-nm (3.0-A) resolution and belong to space group P6(1)22 or its enantiomorph, with cell dimensions a = b = 14.45 nm and c = 16.32 nm. There is one subunit/asymmetric unit which gives a packing density of 2.5 x 10(-3) nm3/Da (2.5 A3/Da), consistent with the experimental crystal density, rho = 1.14 g/cm3. One dimer (approximately 2 x 100 kDa) is located on a crystallographic twofold axis.
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%