Merck
  • Serine protease modulation of Dependence Receptors and EMT protein expression.
Serine protease modulation of Dependence Receptors and EMT protein expression.
Cancer biology & therapy (2018-11-08)
Kara McNair, Caroline M Forrest, Maria C J Vincenten, L Gail Darlington, Trevor W Stone
ABSTRACT
Expression of the tumour suppressor Deleted in Colorectal Cancer (DCC) and the related protein neogenin is reduced by the mammalian serine protease chymotrypsin or the bacterial serine protease subtilisin, with increased cell migration. The present work examines whether these actions are associated with changes in the expression of cadherins, β-catenin and vimentin, established markers of the Epithelial-Mesenchymal Transition (EMT) which has been linked with cell migration and tumour metastasis. The results confirm the depletion of DCC and neogenin and show that chymotrypsin and subtilisin also reduce expression of β-catenin in acutely prepared tissue sections but not in human mammary adenocarcinoma MCF-7 or MDA-MB-231 cells cultured in normal media, or primary normal human breast cells. A loss of β-catenin was also seen in low serum media but transfecting cells with a dcc-containing plasmid induced resistance. E-cadherin was not consistently affected but vimentin was induced by low serum-containing media and was increased by serine proteases in MCF-7 and MDA-MB-231 cells in parallel with increased wound closure. Vimentin might contribute to the promotion of cell migration. The results suggest that changes in EMT proteins depend on the cells or tissues concerned and do not parallel the expression of DCC and neogenin. The increased cell migration induced by serine proteases is not consistently associated with the expression of the EMT proteins implying either that the increased migration may be independent of EMT or supporting the view that EMT is not itself consistently related to migration. (241).
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Nutrient Mixture F-12 Ham, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Human Mammary Epithelial Cell Growth Medium (500ml)
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture