Merck
Defining CRISPR-Cas9 genome-wide nuclease activities with CIRCLE-seq.
Nature protocols (2018-10-21)
Cicera R Lazzarotto, Nhu T Nguyen, Xing Tang, Jose Malagon-Lopez, Jimmy A Guo, Martin J Aryee, J Keith Joung, Shengdar Q Tsai
ABSTRACT
Circularization for in vitro reporting of cleavage effects by sequencing (CIRCLE-seq) is a sensitive and unbiased method for defining the genome-wide activity (on-target and off-target) of CRISPR-Cas9 nucleases by selective sequencing of nuclease-cleaved genomic DNA (gDNA). Here, we describe a detailed experimental and analytical protocol for CIRCLE-seq. The principle of our method is to generate a library of circularized gDNA with minimized numbers of free ends. Highly purified gDNA circles are treated with CRISPR-Cas9 ribonucleoprotein complexes, and nuclease-linearized DNA fragments are then ligated to adapters for high-throughput sequencing. The primary advantages of CIRCLE-seq as compared with other in vitro methods for defining genome-wide genome editing activity are (i) high enrichment for sequencing nuclease-cleaved gDNA/low background, enabling sensitive detection with low sequencing depth requirements; and (ii) the fact that paired-end reads can contain complete information on individual nuclease cleavage sites, enabling use of CIRCLE-seq in species without high-quality reference genomes. The entire protocol can be completed in 2 weeks, including time for gRNA cloning, sequence verification, in vitro transcription, library preparation, and sequencing.
Product Description
Millipore
D-(+)-Glucose monohydrate, suitable for microbiology, ≥99.0%
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
Kanamycin disulfate salt from Streptomyces kanamyceticus, aminoglycoside antibiotic
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%