Merck
Fat & fabulous: bifunctional lipids in the spotlight.
Biochimica et biophysica acta (2014-01-21)
Per Haberkant, Joost C M Holthuis
ABSTRACT
Understanding biological processes at the mechanistic level requires a systematic charting of the physical and functional links between all cellular components. While protein-protein and protein-nucleic acid networks have been subject to many global surveys, other critical cellular components such as membrane lipids have rarely been studied in large-scale interaction screens. Here, we review the development of photoactivatable and clickable lipid analogues-so-called bifunctional lipids-as novel chemical tools that enable a global profiling of lipid-protein interactions in biological membranes. Recent studies indicate that bifunctional lipids hold great promise in systematic efforts to dissect the elaborate crosstalk between proteins and lipids in live cells and organisms. This article is part of a Special Issue entitled Tools to study lipid functions.
Product Description
Avanti
pacFA, Avanti Research - A Croda Brand 900401P, powder
Avanti
pacFA Ceramide, Avanti Research - A Croda Brand 900404P, powder
Avanti
pacFA GlcCer, Avanti Research - A Croda Brand 900405P, powder
Avanti
16:0-pacFA PC, Avanti Research - A Croda Brand 900407P, powder
Avanti
pacFA-18:1 PC, Avanti Research - A Croda Brand 900408P, powder
Avanti
pacFA GalCer, Avanti Research - A Croda Brand 900406P, powder
Sigma-Aldrich
Graphene nanoplatelets, <2 μm particle size
Sigma-Aldrich
SLAP 2,3-Bicyclo-(3,4-THF) N-BnPip Reagent, ≥95%
Sigma-Aldrich
SLAP N-Bn-3Me-Pip Reagent