Merck
Engineering Human Neural Tissue by 3D Bioprinting.
Methods in molecular biology (Clifton, N.J.) (2018-04-22)
Qi Gu, Eva Tomaskovic-Crook, Gordon G Wallace, Jeremy M Crook
ABSTRACT
Bioprinting provides an opportunity to produce three-dimensional (3D) tissues for biomedical research and translational drug discovery, toxicology, and tissue replacement. Here we describe a method for fabricating human neural tissue by 3D printing human neural stem cells with a bioink, and subsequent gelation of the bioink for cell encapsulation, support, and differentiation to functional neurons and supporting neuroglia. The bioink uniquely comprises the polysaccharides alginate, water-soluble carboxymethyl-chitosan, and agarose. Importantly, the method could be adapted to fabricate neural and nonneural tissues from other cell types, with the potential to be applied for both research and clinical product development.
Sigma-Aldrich
ReNcell CX Human Neural Progenitor Cell Line, ReNcell CX is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons & glial cells.
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30