Merck
  • Dramatic enhancement of the detection limits of bioassays via ultrafast deposition of polydopamine.
Dramatic enhancement of the detection limits of bioassays via ultrafast deposition of polydopamine.
Nature biomedical engineering (2017-10-31)
Junwei Li, Madison A Baird, Michael A Davis, Wanyi Tai, Larry S Zweifel, Kristina M Adams Waldorf, Michael Gale, Lakshmi Rajagopal, Robert H Pierce, Xiaohu Gao
ABSTRACT
The ability to detect biomarkers with ultrahigh sensitivity radically transformed biology and disease diagnosis. However, owing to incompatibilities with infrastructure in current biological and medical laboratories, recent innovations in analytical technology have not received broad adoption. Here, we report a simple, universal 'add-on' technology (dubbed EASE) that can be directly plugged into the routine practices of current research and clinical laboratories and that converts the ordinary sensitivities of common bioassays to extraordinary ones. The assay relies on the bioconjugation capabilities and ultrafast and localized deposition of polydopamine at the target site, which permit a large number of reporter molecules to be captured and lead to detection-sensitivity enhancements exceeding 3 orders of magnitude. The application of EASE in the enzyme-linked-immunosorbent-assay-based detection of the HIV antigen in blood from patients leads to a sensitivity lower than 3 fg ml-1. We also show that EASE allows for the direct visualization, in tissues, of the Zika virus and of low-abundance biomarkers related to neurological diseases and cancer immunotherapy.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Lamin A (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution