Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc sulfide
Synonym(s):
Zinc sulphide
Linear Formula:
ZnS
CAS Number:
1314-98-3
Molecular Weight:
97.46
EC Number:
215-251-3
Products
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Physical Form
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
244627
Zinc sulfide,
powder, 10 μm, 99.99% trace metals basis
Expand
View Pricing
333271
Zinc sulfide,
pieces, 3-12 mm, 99.9% trace metals basis
Expand
View Pricing
14459
Zinc sulfide,
purum, 97% (from Zn)
Expand
View Pricing