Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wilmad
®
quick pressure valve NMR tube 5 mm diam.
Products
Product Category
tubes (4)
Manufacturer
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z562866
Wilmad
®
quick pressure valve NMR tube 5 mm diam.,
Medium wall, frequency 300 MHz, L 7 in.
Expand
View Pricing
Z562858
Wilmad
®
quick pressure valve NMR tube 5 mm diam.,
Heavy wall, frequency 300 MHz, L 8 in., pressure -200 psi
Expand
View Pricing
Z562890
Wilmad
®
quick pressure valve NMR tube 5 mm diam.,
Thin wall, frequency 500 MHz, L 8 in.
Expand
View Pricing
Z562815
Wilmad
®
quick pressure valve NMR tube 5 mm diam.,
Thin wall, L 7 in., frequency 300 MHz
Expand
View Pricing