Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wilmad
®
NMR tubes 5 mm diam., precision
Products
Product Category
tubes (30)
Brand
Sigma-Aldrich (7)
Manufacturer
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Package Size
Filter Products
Showing 1-20 of 30
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z272019
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 500 MHz, L 7 in.
Expand
View Pricing
Z272027
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 600 MHz, L 7 in.
Expand
View Pricing
Z274275
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 400 MHz, L 7 in.
Expand
View Pricing
Z271993
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 300 MHz, L 7 in.
Expand
View Pricing
Z274291
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 500 MHz, L 8 in.
Expand
View Pricing
Z274461
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 300 MHz
Expand
View Pricing
Z271985
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L 7 in.
Expand
View Pricing
Z412007
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 800 MHz, L 7 in.
Expand
View Pricing
Z286028
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 200 MHz, L 7 in.
Expand
View Pricing
Z274550
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 600 MHz
Expand
View Pricing
Z684015
Wilmad
®
NMR tubes 5 mm diam., precision,
medium wall, frequency 300 MHz, diam. 5 mm, L 7 in.
Expand
View Pricing
Z274526
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 400 MHz
Expand
View Pricing
Z274445
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L 8 in.
Expand
View Pricing
Z684023
Wilmad
®
NMR tubes 5 mm diam., precision,
medium wall, frequency 400 MHz, diam. 5 mm, L 7 in.
Expand
View Pricing
Z274569
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 600 MHz, L 9 in.
Expand
View Pricing
Z272116
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 300 MHz, L 9 in.
Expand
View Pricing
Z412015
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 800 MHz, L 8 in.
Expand
View Pricing
Z274542
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 500 MHz
Expand
View Pricing
Z684082
Wilmad
®
NMR tubes 5 mm diam., precision,
heavy wall, frequency 300 MHz, diam. 5 mm, L 7 in.
Expand
View Pricing
Z286036
Wilmad
®
NMR tubes 5 mm diam., precision,
frequency 200 MHz, L 8 in.
Expand
View Pricing
Page 1 of 2
Page 1 of 2