Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
WHEATON
®
Type I crimp top serum bottle
Products
Product Category
bottles (15)
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
DWK223748
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (125 mL)
Expand
View Pricing
DWK223760
WHEATON
®
Type I crimp top serum bottle,
without cap, round amber borosilicate glass bottle, capacity (5 mL)
Expand
View Pricing
DWK223746
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (60 mL)
Expand
View Pricing
DWK223763
WHEATON
®
Type I crimp top serum bottle,
without cap, round amber borosilicate glass bottle, capacity (30 mL)
Expand
View Pricing
DWK223761
WHEATON
®
Type I crimp top serum bottle,
without cap, round amber borosilicate glass bottle, capacity (10 mL)
Expand
View Pricing
DWK223764
WHEATON
®
Type I crimp top serum bottle,
without cap, round amber borosilicate glass bottle, capacity (50 mL)
Expand
View Pricing
DWK223766
WHEATON
®
Type I crimp top serum bottle,
without cap, round amber borosilicate glass bottle, capacity (100 mL)
Expand
View Pricing
DWK223743
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (30 mL)
Expand
View Pricing
DWKW223712
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (2 mL)
Expand
View Pricing
DWK223747
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (100 mL)
Expand
View Pricing
DWK223739
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass both filters, capacity (10 mL)
Expand
View Pricing
DWK223762
WHEATON
®
Type I crimp top serum bottle,
without cap, round amber borosilicate glass bottle, capacity (20 mL)
Expand
View Pricing
DWK223745
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (50 mL)
Expand
View Pricing
DWK223738
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (5 mL)
Expand
View Pricing
DWK223742
WHEATON
®
Type I crimp top serum bottle,
without cap, round clear borosilicate glass bottle, capacity (20 mL)
Expand
View Pricing