Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wheaton sample vials with rubber lined caps
Products
Product Category
vials (13)
Brand
Sigma-Aldrich (8)
Supelco (4)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z256064
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 4 mL, thread for 13-425
Expand
View Pricing
Z256161
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 8 mL, thread for 15-425
Expand
View Pricing
Z256056
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 2 mL, thread for 8-425
Expand
View Pricing
Z256218
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 20 mL, thread for 24-400
Expand
View Pricing
Z256234
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 25 mL, thread for 24-400
Expand
View Pricing
Z256188
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 12 mL, thread for 15-425
Expand
View Pricing
Z256242
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 40 mL, thread for 24-400
Expand
View Pricing
Z256226
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 24 mL, thread for 20-400
Expand
View Pricing
Z256285
Wheaton sample vials with rubber lined caps,
amber glass vial, volume 4 mL, thread for 13-425
Expand
View Pricing
Z256196
Wheaton sample vials with rubber lined caps,
clear glass vial, volume 16 mL, thread for 18-400
Expand
View Pricing
Z256293
Wheaton sample vials with rubber lined caps,
amber glass vial, volume 8 mL, thread for 15-425
Expand
View Pricing
Z256269
Wheaton sample vials with rubber lined caps,
clear glass vial (white marking area), volume 8 mL, thread for 15-425
Expand
View Pricing
Z256250
Wheaton sample vials with rubber lined caps,
clear glass vial (white marking area), volume 4 mL, thread for 13-425
Expand
View Pricing