Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wheaton sample vials
Products
Product Category
vials (7)
Brand
Sigma-Aldrich (2)
Supelco (5)
Color
Diameter (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z188700
Wheaton sample vials,
clear, volume 4 mL, cap size 13 - 425, diam. × H 15 mm × 46 mm
Expand
View Pricing
Z188727
Wheaton sample vials,
clear, volume 12 mL, cap size 15 - 425, diam. × H 19 mm × 66 mm
Expand
View Pricing
Z188697
Wheaton sample vials,
clear, volume 2 mL, cap size 8 - 425, diam. × H 12 mm × 36 mm
Expand
View Pricing
Z188719
Wheaton sample vials,
clear, volume 8 mL, cap size 15 - 425, diam. × H 17 mm × 61 mm
Expand
View Pricing
Z188743
Wheaton sample vials,
clear, volume 24 mL, cap size 20 - 400, diam. × H 23 mm × 86 mm
Expand
View Pricing
Z188786
Wheaton sample vials,
amber, volume 4 mL, cap size 13 - 425, diam. × H 15 mm × 46 mm
Expand
View Pricing
Z188794
Wheaton sample vials,
amber, volume 8 mL, cap size 15 - 425, diam. × H 17 mm × 61 mm
Expand
View Pricing