Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wheaton glass serum bottle
Products
Product Category
bottles (9)
Brand
Sigma-Aldrich (9)
Color
Capacity (mL)
Diameter (mm)
Height (mm)
OD (mm)
Volume (ml)
Closure Type
Bottle material
Vial Bottle Material
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z114014
Wheaton glass serum bottle,
volume 125 mL, clear, O.D. 20 mm, diam. × H 54 mm × 107 mm
Expand
View Pricing
Z113999
Wheaton glass serum bottle,
volume 50 mL, clear, O.D. 20 mm, diam. × H 43 mm × 73 mm
Expand
View Pricing
Z114006
Wheaton glass serum bottle,
volume 100 mL, clear, O.D. 20 mm, diam. × H 52 mm × 95 mm
Expand
View Pricing
Z114049
Wheaton glass serum bottle,
volume 30 mL, amber, O.D. 20 mm, diam. × H 37 mm × 63 mm
Expand
View Pricing
Z113980
Wheaton glass serum bottle,
volume 30 mL, clear, O.D. 20 mm, diam. × H 37 mm × 63 mm
Expand
View Pricing
Z114057
Wheaton glass serum bottle,
volume 50 mL, amber, O.D. 20 mm, diam. × H 43 mm × 73 mm
Expand
View Pricing
Z113972
Wheaton glass serum bottle,
volume 10 mL, clear, O.D. 20 mm, diam. × H 25 mm × 54 mm
Expand
View Pricing
Z113964
Wheaton glass serum bottle,
volume 5 mL, clear, O.D. 20 mm, diam. × H 23 mm × 47 mm
Expand
View Pricing
Z114065
Wheaton glass serum bottle,
volume 100 mL, amber, O.D. 20 mm, diam. × H 52 mm × 95 mm
Expand
View Pricing