Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Uniflo
®
13 syringe filters
Products
Product Category
syringe filters (10)
Manufacturer
Diameter (mm)
Surface Area cm²
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
WHA9916-1302
Whatman
®
Uniflo
®
13 syringe filters,
polyethersulfone membrane, sterile, pore size 0.2 μm, pack of 100 ea
Expand
View Pricing
WHA9916-1304
Whatman
®
Uniflo
®
13 syringe filters,
polyethersulfone membrane, sterile, pore size 0.45 μm, pack of 100 ea
Expand
View Pricing
WHA99111302
Whatman
®
Uniflo
®
13 syringe filters,
PTFE membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA99101302
Whatman
®
Uniflo
®
13 syringe filters,
Nylon membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA99091302
Whatman
®
Uniflo
®
13 syringe filters,
PVDF membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA99091304
Whatman
®
Uniflo
®
13 syringe filters,
PVDF membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA99111304
Whatman
®
Uniflo
®
13 syringe filters,
PTFE membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA99101304
Whatman
®
Uniflo
®
13 syringe filters,
Nylon membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA99121304
Whatman
®
Uniflo
®
13 syringe filters,
polyethersulfone membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA99121302
Whatman
®
Uniflo
®
13 syringe filters,
polyethersulfone membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing