Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
qualitative filter paper, Grade 597
Products
Product Category
filter discs (13)
Manufacturer
Diameter (mm)
Package Size
Pore size (µm)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
WHA10311809
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 90 mm, pack of 100
Expand
View Pricing
WHA10311810
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 110 mm, pack of 100
Expand
View Pricing
WHA10311804
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 45 mm, pack of 100
Expand
View Pricing
WHA10311814
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 185 mm, pack of 100
Expand
View Pricing
WHA10311811
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 125 mm, pack of 100
Expand
View Pricing
WHA10311807
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 55 mm, pack of 100
Expand
View Pricing
WHA10311862
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 12.7 mm, pack of 100
Expand
View Pricing
WHA10311897
Whatman
®
qualitative filter paper, Grade 597,
sheets, W × L 580 mm × 580 mm, pack of 100
Expand
View Pricing
WHA10311887
Whatman
®
qualitative filter paper, Grade 597,
sheets, L × W 580 mm × 580 mm, pack of 500
Expand
View Pricing
WHA10311808
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 70 mm, pack of 100
Expand
View Pricing
WHA10311820
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 240 mm, pack of 100
Expand
View Pricing
WHA10311812
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 150 mm, pack of 100
Expand
View Pricing
WHA98949552
Whatman
®
qualitative filter paper, Grade 597,
grade, 597, cellulose filter, filter diam. 55 mm, non-sterile, case of 100 ea
Expand
View Pricing
WHA10311822
Whatman
®
qualitative filter paper, Grade 597,
circles, diam. 320 mm, pack of 100
Expand
View Pricing