Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
qualitative filter paper, Grade 4
Products
Product Category
filter discs (18)
Manufacturer
Diameter (mm)
Package Size
Pore size (µm)
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
WHA1004125
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 125 mm, pack of 100
Expand
View Pricing
WHA1004150
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 150 mm, pack of 100
Expand
View Pricing
WHA1004110
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 110 mm, pack of 100
Expand
View Pricing
WHA1004070
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 70 mm, pack of 100
Expand
View Pricing
WHA1004090
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 90 mm, pack of 100
Expand
View Pricing
WHA1004047
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 47 mm, pack of 100
Expand
View Pricing
WHA1004185
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 185 mm, pack of 100
Expand
View Pricing
WHA1004240
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 240 mm, pack of 100
Expand
View Pricing
WHA1004055
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 55 mm, pack of 100
Expand
View Pricing
WHA1004042
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 42.5 mm, pack of 100
Expand
View Pricing
WHA1004050
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 50 mm, pack of 100
Expand
View Pricing
WHA1004648
Whatman
®
qualitative filter paper, Grade 4,
roll, W × L 38 mm × 114 mm, pack of 1
Expand
View Pricing
WHA1004917
Whatman
®
qualitative filter paper, Grade 4,
sheets, W × L 460 mm × 570 mm, pack of 100
Expand
View Pricing
WHA1004400
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 400 mm, pack of 100
Expand
View Pricing
WHA1004930
Whatman
®
qualitative filter paper, Grade 4,
sheets, W × L 580 mm × 580, pack of 100
Expand
View Pricing
WHA1004320
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 320 mm, pack of 100
Expand
View Pricing
WHA1004270
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 270 mm, pack of 100
Expand
View Pricing
WHA1004027
Whatman
®
qualitative filter paper, Grade 4,
circles, diam. 27 mm, pack of 400
Expand
View Pricing