Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
quadrant folded cellulose fiber filter papers
Products
Manufacturer
Diameter (mm)
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
WHA10380405
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 1 FF, diam. (125 mm), pack of 500 ea
Expand
View Pricing
WHA10380404
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 1 FF, diam. (110 mm), pack of 500 ea
Expand
View Pricing
WHA10380206
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 41 FF, diam. (150 mm), pack of 500 ea
Expand
View Pricing
WHA10380205
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 41 FF, diam. (125 mm), pack of 500 ea
Expand
View Pricing
WHA10380406
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 1 FF, diam. (150 mm), pack of 500 ea
Expand
View Pricing
WHA10380004
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 40 FF, diam. (110 mm), pack of 500 ea
Expand
View Pricing
WHA10380005
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 40 FF, diam. (125 mm), pack of 500 ea
Expand
View Pricing
WHA10380006
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 40 FF, diam. (150 mm), pack of 500 ea
Expand
View Pricing
WHA10380204
Whatman
®
quadrant folded cellulose fiber filter papers,
Grade 41 FF, diam. (110 mm), pack of 500 ea
Expand
View Pricing