<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V
Products
Product Category
filter discs (6)
Manufacturer
Diameter (mm)
Package Size
Pore size (µm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
WHA1201185
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V,
circles, diam. 185 mm, pack of 100
Expand
View Pricing
WHA1201320
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V,
circles, diam. 32 cm, pack of 100
Expand
View Pricing
WHA1201150
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V,
circles, diam. 15 cm, pack of 100
Expand
View Pricing
WHA1201240
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V,
circles, diam. 24 cm, pack of 100
Expand
View Pricing
WHA1201125
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V,
circles, diam. 125 mm, pack of 100
Expand
View Pricing
WHA1201270
Whatman
®
Prepleated Qualitative Filter Paper, Grade 1V,
circles, diam. 270 mm, pack of 100
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More