Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
prepleated filter paper, Grade 802, creped
Products
Product Category
filter discs (6)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
WHA5802320
Whatman
®
prepleated filter paper, Grade 802, creped,
circles, diam. 320 mm (12.6 in.), pkg of 100 ea
Expand
View Pricing
WHA5802185
Whatman
®
prepleated filter paper, Grade 802, creped,
circles, diam. 185 mm (7.3 in.), pkg of 100 ea
Expand
View Pricing
WHA5802150
Whatman
®
prepleated filter paper, Grade 802, creped,
circles, diam. 150 mm (5.9 in.), pkg of 100 ea
Expand
View Pricing
WHA5802240
Whatman
®
prepleated filter paper, Grade 802, creped,
circles, diam. 240 mm (9.4 in.), pkg of 100 ea
Expand
View Pricing
WHA5802125
Whatman
®
prepleated filter paper, Grade 802, creped,
circles, diam. 125 mm (4.9 in.), pkg of 100 ea
Expand
View Pricing
WHA5802385
Whatman
®
prepleated filter paper, Grade 802, creped,
circles, diam. 385 mm (15.2 in.), pkg of 100 ea
Expand
View Pricing