Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size
Products
Product Category
filter discs (7)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
WHA7182004
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 100 ea, 47 mm diam
Expand
View Pricing
WHA7182009
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 25 ea, 90 mm diam
Expand
View Pricing
WHA7182002
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 100 ea, 25 mm diam
Expand
View Pricing
WHA10401312
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 100 ea, 47 mm diam
Expand
View Pricing
WHA10401314
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 100 ea, 50 mm diam
Expand
View Pricing
WHA7182001
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 100 ea, 13 mm diam
Expand
View Pricing
WHA7182014
Whatman
®
nitrocellulose NC20 filter discs 0.2 μm pore size,
white, hydrophilic, 25 ea, 142mm diam
Expand
View Pricing