Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
multiple vacuum filtration device
Products
Product Category
holders (8)
Manufacturer
Capacity (mL)
Housing Material
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
WHA10498762
Whatman
®
multiple vacuum filtration device,
6-place, 6-place manifold, stainless steel
Expand
View Pricing
WHA10498761
Whatman
®
multiple vacuum filtration device,
3-place, 3-place manifold, stainless steel
Expand
View Pricing
WHA10445850
Whatman
®
multiple vacuum filtration device,
3-place, AS300/5, stainless steel, capacity 100 mL, 47/50, support screen
Expand
View Pricing
WHA10445830
Whatman
®
multiple vacuum filtration device,
3-place, AS300/3, stainless steel, capacity 500 mL, 47/50, support screen
Expand
View Pricing
WHA10445835
Whatman
®
multiple vacuum filtration device,
3-place, AS310/3, stainless steel, capacity 500 mL, 47/50, support screen
Expand
View Pricing
WHA10444850
Whatman
®
multiple vacuum filtration device,
6-place, AS600/5, stainless steel, capacity 100 mL, 47/50, support screen
Expand
View Pricing
WHA10444835
Whatman
®
multiple vacuum filtration device,
6-place, AS610/3, stainless steel, capacity 500 mL, 47/50, support screen
Expand
View Pricing
WHA10444830
Whatman
®
multiple vacuum filtration device,
6-place, AS600/3, stainless steel, capacity 500 mL, 47/50, support screen
Expand
View Pricing