Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Mini-UniPrep
®
G2 slit septum
Products
Product Category
filters (8)
Manufacturer
Cap
Color
Housing Material
Membrane Material
Vial Bottle Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GS203NPEORG
Whatman
®
Mini-UniPrep
®
G2 slit septum,
PTFE membrane, pore size 0.2 μm, clear vial, pack of 100
Expand
View Pricing
GS203NPUORG
Whatman
®
Mini-UniPrep
®
G2 slit septum,
PTFE membrane, pore size 0.45 μm, clear vial, pack of 100
Expand
View Pricing
GS203NPUGMF
Whatman
®
Mini-UniPrep
®
G2 slit septum,
glass microfiber membrane, pore size 0.45 μm, clear vial, pack of 100
Expand
View Pricing
GS203NPENYL
Whatman
®
Mini-UniPrep
®
G2 slit septum,
Nylon membrane, pore size 0.2 μm, clear vial, pack of 100
Expand
View Pricing
GS203NPUAQU
Whatman
®
Mini-UniPrep
®
G2 slit septum,
PVDF membrane, pore size 0.45 μm, clear vial, pack of 100
Expand
View Pricing
GS503NPEORG
Whatman
®
Mini-UniPrep
®
G2 slit septum,
PTFE membrane, pore size 0.2 μm, clear vial, pack of 1000
Expand
View Pricing
GS203NPEAQU
Whatman
®
Mini-UniPrep
®
G2 slit septum,
PVDF membrane, pore size 0.2 μm, clear vial, pack of 100
Expand
View Pricing
GS503NPUORG
Whatman
®
Mini-UniPrep
®
G2 slit septum,
PTFE membrane, pore size 0.45 μm, clear vial, pack of 1000
Expand
View Pricing