Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Whatman
®
glass vacuum filtration device
Products
Product Category
holders (28)
filters (2)
Manufacturer
Capacity (mL)
Filter Capacity (ml)
Housing Material
Package Size
Sterility
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
WHA10441001
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/0/1 funnel, capacity 60 mL
Expand
View Pricing
WHA10441004
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/0/05 PTFE centering ring
Expand
View Pricing
WHA10441003
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/0/03 glass frit, diam. 25 mm
Expand
View Pricing
WHA10442200
Whatman
®
glass vacuum filtration device,
Expand
View Pricing
WHA10442300
Whatman
®
glass vacuum filtration device,
GV050 series, GV050/3 vacuum filter holder & flask, 250 mL, 47/50 mm, sieve, 1000 mL Erlenmeyer
Expand
View Pricing
WHA10441203
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/2/08 Erlenmeyer flask
Expand
View Pricing
WHA10441005
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/0/06 spring-action clamp
Expand
View Pricing
WHA10442000
Whatman
®
glass vacuum filtration device,
GV 050/0, glass frit filter, rubber stopper connection, 1/pk
Expand
View Pricing
WHA10442203
Whatman
®
glass vacuum filtration device,
GV050 series, GV050/2/08 Erlenmeyer flask, NS29, 1000 mL
Expand
View Pricing
WHA10441200
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/2 vacuum filter holder & flask
Expand
View Pricing
WHA10441201
Whatman
®
glass vacuum filtration device,
GV025 series, GV025/2/02 hose connector RD14
Expand
View Pricing
WHA10442100
Whatman
®
glass vacuum filtration device,
GV 050/1, sieve filter, rubber stopper connection, 1/pk
Expand
View Pricing
WHA10442213
Whatman
®
glass vacuum filtration device,
GV050 series, GV050/2/13 Erlenmeyer flask, NS45, 1000 mL
Expand
View Pricing
WHA10442001
Whatman
®
glass vacuum filtration device,
GV050/0/01, funnel without cap, 250 ml, 1/pk
Expand
View Pricing
WHA10443001
Whatman
®
glass vacuum filtration device,
GV100 series, GV100/0/01 funnel, 500 mL
Expand
View Pricing
WHA10442004
Whatman
®
glass vacuum filtration device,
GV050 series, GV050/0/05 PTFE centering ring to be used with GV050/0/03 glass frit
Expand
View Pricing
WHA10443003
Whatman
®
glass vacuum filtration device,
GV100 series, GV100/0/03 glass frit
Expand
View Pricing
WHA10442212
Whatman
®
glass vacuum filtration device,
GV050/2/12, lower part, glass, NS45, for GV050/2, 1/pk
Expand
View Pricing
WHA10443000
Whatman
®
glass vacuum filtration device,
GV100 series, GV100/0 vacuum filter holder, 250 mL, 100 mm, glass frit
Expand
View Pricing
WHA10442201
Whatman
®
glass vacuum filtration device,
GV050 series, GV050/2/02 lower part for GV050/02, NS29
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
Page 1 of 2
Page 1 of 2
English - EN
Learn More