Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size
Products
Product Category
filter discs (6)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
WHA10407112
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size,
white, hydrophilic, black grid (3.1 mm), sterile, for membrane butler, 400 ea, 47 mm diam
Expand
View Pricing
WHA10407132
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size,
black, hydrophilic, white grid (3.1 mm), sterile, for membrane butler, 400 ea, 47 mm diam
Expand
View Pricing
WHA10407114
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size,
white, hydrophilic, black grid (3.1 mm), sterile, for membrane butler, 400 ea, 50 mm diam
Expand
View Pricing
WHA10407170
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size,
green, hydrophilic, black grid (3.1 mm), sterile, for membrane butler, 400 ea, 47 mm diam
Expand
View Pricing
WHA10407172
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size,
green, hydrophilic, black grid (3.1 mm), sterile, for membrane butler, 400 ea, 50 mm diam
Expand
View Pricing
WHA10407134
Whatman
®
celluose nitrate MicroPlus-21 STL filter discs 0.45 μm pore size,
black, hydrophilic, white grid (3.1 mm), sterile, for membrane butler, 400 ea, 50 mm diam
Expand
View Pricing