Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, screw top, amber glass (vial only)
Products
Product Category
vials (14)
Brand
Supelco (13)
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
27083-U
Vials, screw top, amber glass (vial only),
volume 2 mL, amber glass vial, thread for 8-425, pkg of 100 ea
Expand
View Pricing
27088-U
Vials, screw top, amber glass (vial only),
volume 15 mL, amber glass vial, thread for 18-400, pkg of 100 ea
Expand
View Pricing
27115-U
Vials, screw top, amber glass (vial only),
volume 4 mL, amber glass vial, thread for 13-425, pkg of 100 ea
Expand
View Pricing
27072-U
Vials, screw top, amber glass (vial only),
volume 7 mL, amber glass vial, thread for 15-425, pkg of 100 ea
Expand
View Pricing
27267-U
Vials, screw top, amber glass (vial only),
volume 2 mL, amber glass vial, thread for 10-425, pkg of 100 ea
Expand
View Pricing
27382
Vials, screw top, amber glass (vial only),
volume 40 mL, amber glass vial, thread for 24-400, pkg of 100 ea
Expand
View Pricing
27073-U
Vials, screw top, amber glass (vial only),
volume 22 mL, amber glass vial, thread for 20-400, pkg of 100 ea
Expand
View Pricing
27032
Vials, screw top, amber glass (vial only),
volume 4 mL, amber glass vial, thread for 13-425, pkg of 1000 ea
Expand
View Pricing
854986
Vials, screw top, amber glass (vial only),
volume 4 mL, amber glass vial, thread for 13-425, pkg of 100 ea
Expand
View Pricing
507628
Vials, screw top, amber glass (vial only),
volume 2 mL, amber glass vial, thread for 8-425, pkg of 100 ea
Expand
View Pricing
27185-U
Vials, screw top, amber glass (vial only),
volume 40 mL, amber glass vial, thread for 24-400, pkg of 100 ea
Expand
View Pricing
23702-U
Vials, screw top, amber glass (vial only),
volume 40 mL, amber glass vial, thread for 24-400 24-400, pkg of 100 ea
Expand
View Pricing
27116-U
Vials, screw top, amber glass (vial only),
volume 4 mL, amber glass vial (with marking spot), thread for 13-425, pkg of 100 ea
Expand
View Pricing
27085-U
Vials, screw top, amber glass (vial only),
volume 2 mL, amber glass vial (with marking spot), thread for 8-425, pkg of 1000 ea
Expand
View Pricing