Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, pre-cleaned and assembled, pk of 100
Products
Product Category
vials (10)
Brand
Supelco (10)
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
27340
Vials, pre-cleaned and assembled, pk of 100,
volume 4 mL, clear glass vial
Expand
View Pricing
27345
Vials, pre-cleaned and assembled, pk of 100,
volume 4 mL, amber glass vial
Expand
View Pricing
27348
Vials, pre-cleaned and assembled, pk of 100,
volume 22 mL, amber glass vial
Expand
View Pricing
27342
Vials, pre-cleaned and assembled, pk of 100,
volume 15 mL, clear glass vial
Expand
View Pricing
27346
Vials, pre-cleaned and assembled, pk of 100,
volume 7 mL, amber glass vial
Expand
View Pricing
27344
Vials, pre-cleaned and assembled, pk of 100,
volume 2 mL, amber glass vial
Expand
View Pricing
27339
Vials, pre-cleaned and assembled, pk of 100,
volume 2 mL, clear glass vial
Expand
View Pricing
27341
Vials, pre-cleaned and assembled, pk of 100,
volume 7 mL, clear glass vial
Expand
View Pricing
27343
Vials, pre-cleaned and assembled, pk of 100,
volume 22 mL, clear glass vial
Expand
View Pricing
27347
Vials, pre-cleaned and assembled, pk of 100,
volume 15 mL, amber glass vial
Expand
View Pricing