Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, 2 mL crimp top, clear glass, 12 x 32 mm
Products
Product Category
vials (4)
Brand
Supelco (4)
Color
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
27058
Vials, 2 mL crimp top, clear glass, 12 x 32 mm,
clear glass vial (large opening), pkg of 100 ea
Expand
View Pricing
27063
Vials, 2 mL crimp top, clear glass, 12 x 32 mm,
clear glass vial (with marking spot), pkg of 1000 ea
Expand
View Pricing
27059
Vials, 2 mL crimp top, clear glass, 12 x 32 mm,
clear glass vial (large opening)
Expand
View Pricing
27069
Vials, 2 mL crimp top, clear glass, 12 x 32 mm,
volume 2 mL, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, pkg of 1000 ea
Expand
View Pricing