Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Veratridine
Synonym(s):
3-Veratroylveracevine, Veracevine veratrate, Veratridine
Empirical Formula (Hill Notation):
C
36
H
51
NO
11
CAS Number:
71-62-5
Molecular Weight:
673.79
EC Number:
200-758-4
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
V5754
Veratridine,
≥90% (HPLC), powder, Na+ channel opener
Expand
View Pricing
676950-M
Veratridine,
≥98% (TLC), solid, Na⁺ channel opener, Calbiochem
Expand
View Pricing
PHL82325
Veratridine,
phyproof
®
Reference Substance
Expand
View Pricing