Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
V-bottom assay plate
Products
Product Category
plates (4)
Color
Manufacturer
Binding Type
Wells
Color
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
AXYP96450VCS
V-bottom assay plate,
size 96 wells, V-bottom clear polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs, sterile
Expand
View Pricing
AXYP96450VC
V-bottom assay plate,
size 96 wells, maximum volume 500 μL, V-bottom clear polypropylene wells, pkg of 10x5plates/cs
Expand
View Pricing
AXYP96450VB
V-bottom assay plate,
size 96 wells, V-bottom blue polypropylene wells, SBS FootPrint, maximum volume 500 μL, blue, pkg of 10x5plates/cs
Expand
View Pricing
AXYP96450VY
V-bottom assay plate,
size 96 wells, SBS FootPrint, V-bottom yellow polypropylene wells, yellow, maximum volume 500 μL, pkg of 10x5plates/cs
Expand
View Pricing