Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Umbelliferone
Synonym(s):
7-Hydroxy-2
H
-1-benzopyran-2-one, 7-Hydroxycoumarin
Empirical Formula (Hill Notation):
C
9
H
6
O
3
CAS Number:
93-35-6
Molecular Weight:
162.14
EC Number:
202-240-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (2)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
H24003
Umbelliferone,
99%
Expand
View Pricing
93979
Umbelliferone,
suitable for fluorescence indicator, ≥98.0% (HPLC)
Expand
View Pricing
PHL80098
Umbelliferone,
phyproof
®
Reference Substance
Expand
View Pricing
54826
Umbelliferone,
analytical standard
Expand
View Pricing