Skip to Content
Merck
Tungsten(VI) oxychloride 98%

Tungsten(VI) oxychloride

Tungsten(VI) oxychloride 98%
Synonym(s):
Tungsten(VI) chloride oxide, Tungsten(VI) oxide chloride
Linear Formula:
WOCl4
CAS Number:
Molecular Weight:
341.65
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing