Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten-copper alloy, W60Cu40
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF58108226
Tungsten-copper alloy, W60Cu40,
185mm wire, 0.5mm diameter, straight wire
Expand
View Pricing
GF22072890
Tungsten-copper alloy, W60Cu40,
foil, 25 x 50mm, 0.25mm thickness
Expand
View Pricing
GF27331837
Tungsten-copper alloy, W60Cu40,
foil, 25 x 25mm, 0.25mm thickness
Expand
View Pricing
GF93881560
Tungsten-copper alloy, W60Cu40,
foil, 25 x 50mm, 0.125mm thickness
Expand
View Pricing
GF98272626
Tungsten-copper alloy, W60Cu40,
foil, 25 x 75mm, 1.0mm thickness
Expand
View Pricing
GF35195014
Tungsten-copper alloy, W60Cu40,
foil, 25 x 100mm, 0.125mm thickness
Expand
View Pricing
GF70512331
Tungsten-copper alloy, W60Cu40,
175mm tube, 0.5mm outside diameter, 0.13mm wall thickness, 0.24mm inside diameter
Expand
View Pricing
GF55219484
Tungsten-copper alloy, W60Cu40,
175mm tube, 3.15mm outside diameter, 1.15mm wall thickness, 0.85mm inside diameter
Expand
View Pricing
GF89483158
Tungsten-copper alloy, W60Cu40,
80mm tube, 3.15mm outside diameter, 1.15mm wall thickness, 0.85mm inside diameter
Expand
View Pricing
GF09716216
Tungsten-copper alloy, W60Cu40,
foil, 75 x 100mm, 1.0mm thickness
Expand
View Pricing
GF84077725
Tungsten-copper alloy, W60Cu40,
foil, 50 x 75mm, 1.0mm thickness
Expand
View Pricing
GF98410383
Tungsten-copper alloy, W60Cu40,
foil, 25 x 75mm, 0.50mm thickness
Expand
View Pricing
GF10742420
Tungsten-copper alloy, W60Cu40,
foil, 25 x 75mm, 0.25mm thickness
Expand
View Pricing
GF62351052
Tungsten-copper alloy, W60Cu40,
foil, 25 x 25mm, 0.125mm thickness
Expand
View Pricing
GF62188689
Tungsten-copper alloy, W60Cu40,
foil, 75 x 100mm, 0.50mm thickness
Expand
View Pricing
GF01297504
Tungsten-copper alloy, W60Cu40,
foil, 50 x 75mm, 0.50mm thickness
Expand
View Pricing
GF90572878
Tungsten-copper alloy, W60Cu40,
foil, 75 x 100mm, 0.25mm thickness
Expand
View Pricing
GF96093085
Tungsten-copper alloy, W60Cu40,
foil, 50 x 75mm, 0.25mm thickness
Expand
View Pricing
GF99376587
Tungsten-copper alloy, W60Cu40,
foil, 25 x 200mm, 0.125mm thickness
Expand
View Pricing
GF71820783
Tungsten-copper alloy, W60Cu40,
foil, 75 x 200mm, 0.25mm thickness
Expand
View Pricing
Page 1 of 2
Page 1 of 2