Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tunair
™
shake flask
Products
Product Category
flasks (6)
Manufacturer
Binding Type
Volume (ml)
Working Volume (ml)
Cap
Closure Type
Color
Package Size
Sterility
Style
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z710822
Tunair
™
shake flask,
flask volume 2,500 mL, clear polypropylene, full baffle
Expand
View Pricing
Z710814
Tunair
™
shake flask,
flask volume 2,500 mL, polypropylene, half baffle
Expand
View Pricing
Z710792
Tunair
™
shake flask,
flask volume 300 mL, wide-mouth polypropylene flask, full baffle
Expand
View Pricing
Z710784
Tunair
™
shake flask,
flask volume 300 mL, polypropylene flask, half baffle
Expand
View Pricing
Z710776
Tunair
™
shake flask,
flask volume 300 mL, polypropylene flask, no baffle
Expand
View Pricing
Z710806
Tunair
™
shake flask,
flask volume 2,500 mL, polypropylene, no baffle
Expand
View Pricing