Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tris(2-chloroethyl) phosphate
Synonym(s):
Phosphoric acid tris(2-chloroethyl) ester
Linear Formula:
(ClCH
2
CH
2
O)
3
P(O)
CAS Number:
115-96-8
Molecular Weight:
285.49
EC Number:
204-118-5
Products
Product Category
building blocks (1)
plasticizer (1)
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
119660
Tris(2-chloroethyl) phosphate,
97%
Expand
View Pricing
96382
Tris(2-chloroethyl) phosphate,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing