Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trimethylolpropane ethoxylate triacrylate
Linear Formula:
[H
2
C=CHCO
2
(CH
2
CH
2
O)
n
CH
2
]
3
CC
2
H
5
CAS Number:
28961-43-5
Products
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Physical Form
Monomer Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
409073
Trimethylolpropane ethoxylate triacrylate,
average M
n
~428
Expand
View Pricing
412198
Trimethylolpropane ethoxylate triacrylate,
average M
n
~912
Expand
View Pricing