Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trimethoxy[2-(7-oxabicyclo[4.1.0]hept-3-yl)ethyl]silane
Synonym(s):
[2-(3,4-Epoxycyclohexyl)ethyl]trimethoxysilane, 2-(7-Oxabicyclo[4.1.0]hept-3-yl)ethyl-trimethoxysilane, 3-(2-Trimethoxysilylethyl)cyclohexene oxide
Empirical Formula (Hill Notation):
C
11
H
22
O
4
Si
CAS Number:
3388-04-3
Molecular Weight:
246.38
EC Number:
222-217-1
Products
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
413321
Trimethoxy[2-(7-oxabicyclo[4.1.0]hept-3-yl)ethyl]silane,
98%
Expand
View Pricing