Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Transfer pipette, polyethylene
Products
Product Category
pipettes (53)
Brand
Sigma-Aldrich (33)
Manufacturer
Diameter (mm)
Length (mm)
Package Size
Sterility
Filter Products
Showing 1-20 of 53
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
Z135003
Transfer pipette, polyethylene,
general purpose, standard, bulb draw 3.2 mL, non-sterile
Expand
View Pricing
Z350605
Transfer pipette, polyethylene,
general purpose, standard, bulb draw 3.2 mL, sterile
Expand
View Pricing
Z350796
Transfer pipette, polyethylene,
graduated to 3 mL, bulb draw 3.2 mL, non-sterile
Expand
View Pricing
Z135011
Transfer pipette, polyethylene,
graduated to 1 mL, bulb draw 3.1 mL, non-sterile
Expand
View Pricing
Z135089
Transfer pipette, polyethylene,
graduated to 1 mL, bulb draw 5.0 mL, sterile
Expand
View Pricing
Z354333
Transfer pipette, polyethylene,
fine tip extended, small bulb, draw 1.0 mL, non-sterile
Expand
View Pricing
Z350753
Transfer pipette, polyethylene,
general purpose, extra long (12"), bulb draw 7.1 mL, non-sterile
Expand
View Pricing
Z350761
Transfer pipette, polyethylene,
general purpose, extra long (12"), bulb draw 7.1 mL, sterile
Expand
View Pricing
Z350613
Transfer pipette, polyethylene,
general purpose, large bulb, draw 4.6 mL, non-sterile
Expand
View Pricing
Z350958
Transfer pipette, polyethylene,
fine tip, short stem, bulb draw 1.0 mL, non-sterile
Expand
View Pricing
Z350656
Transfer pipette, polyethylene,
general purpose, short, bulb draw 0.9 mL, non-sterile
Expand
View Pricing
Z350826
Transfer pipette, polyethylene,
graduated to 3 mL, bulb draw 3.2 mL, sterile
Expand
View Pricing
Z350915
Transfer pipette, polyethylene,
fine tip extended, small bulb, draw 1.0 mL, sterile
Expand
View Pricing
Z135054
Transfer pipette, polyethylene,
general purpose, long (9"), bulb draw 2.3 mL, non-sterile
Expand
View Pricing
Z354309
Transfer pipette, polyethylene,
graduated to 2 mL, blood bank, bulb draw 1.8 mL, non-sterile
Expand
View Pricing
Z135046
Transfer pipette, polyethylene,
narrow standard stem, bulb draw 3.1 mL, non-sterile
Expand
View Pricing
Z350818
Transfer pipette, polyethylene,
graduated to 3 mL, bulb draw 3.2 mL, sterile
Expand
View Pricing
Z354368
Transfer pipette, polyethylene,
fine tip extended, large bulb, draw 3.0 mL, non-sterile
Expand
View Pricing
Z135038
Transfer pipette, polyethylene,
fine tip standard, bulb draw 3.3 mL, non-sterile
Expand
View Pricing
Z350923
Transfer pipette, polyethylene,
fine tip extended, large bulb, draw 3.0 mL, sterile
Expand
View Pricing
Page 1 of 3
Page 1 of 3