Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TPP
®
tissue culture plates
Products
Product Category
plates (10)
Manufacturer
Binding Type
Height (mm)
Length (mm)
Surface Area cm²
Volume (ml)
Wells
Working Volume (ml)
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z707902
TPP
®
tissue culture plates,
96 well plate, flat bottom, polystyrene, 0.34cm
2
, sterile, 108/cs
Expand
View Pricing
Z707791
TPP
®
tissue culture plates,
24 well plate, flat bottom, polystyrene, 1.86 cm
2
,
sterile, 126/cs
Expand
View Pricing
Z707767
TPP
®
tissue culture plates,
6 well plate, flat bottom, polystyrene, 8.96 cm
2
,
sterile, 64/cs
Expand
View Pricing
Z707759
TPP
®
tissue culture plates,
6 well plate, flat bottom, polystyrene
,
sterile, 126/cs
Expand
View Pricing
Z707910
TPP
®
tissue culture plates,
Expand
View Pricing
Z707805
TPP
®
tissue culture plates,
24 well plate, flat bottom, polystyrene, 1.86 cm
2
,
sterile, 72/cs
Expand
View Pricing
Z707775
TPP
®
tissue culture plates,
12 well plate, flat bottom, polystyrene, 3.60 cm
2
,
sterile, 126/cs
Expand
View Pricing
Z707899
TPP
®
tissue culture plates,
96 well plate, round bottom, polystyrene, 0.40cm
2
, sterile, 162/cs
Expand
View Pricing
Z707783
TPP
®
tissue culture plates,
12 well plate, flat bottom, polystyrene, 3.6 cm
2
,
sterile, 72/cs
Expand
View Pricing
Z707880
TPP
®
tissue culture plates,
96 well plate, round bottom, polystyrene
,
sterile, 108/cs
Expand
View Pricing