Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Total Acid Number Standard
Synonym(s):
TAN-Standard
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
TAN005
Total Acid Number Standard,
ASTM D664 (0.49 mg KOH/g), pkg of 1 × 125 g
Expand
View Pricing
TAN001
Total Acid Number Standard,
ASTM D664 (0.10 mg KOH/g), pkg of 1 × 125 g
Expand
View Pricing
TAN015
Total Acid Number Standard,
ASTM D664 (1.54 mg KOH/g), pkg of 1 × 125 g
Expand
View Pricing
TAN030
Total Acid Number Standard,
ASTM D664 (2.93 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TAN050
Total Acid Number Standard,
ASTM D664 (4.57 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing
TAN025
Total Acid Number Standard,
ASTM D664 (2.48 mg KOH/g), pkg of 1 × 50 g
Expand
View Pricing