Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium(IV) oxide, rutile
Synonym(s):
Titanium dioxide
Linear Formula:
TiO
2
CAS Number:
1317-80-2
Molecular Weight:
79.87
EC Number:
215-282-2
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Length (mm)
Application
Particle Size (µm)
Reagent Type
Core
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
224227
Titanium(IV) oxide, rutile,
powder, <5 μm, ≥99.9% trace metals basis
Expand
View Pricing
637262
Titanium(IV) oxide, rutile,
nanopowder, <100 nm particle size, 99.5% trace metals basis
Expand
View Pricing
204757
Titanium(IV) oxide, rutile,
≥99.98% trace metals basis
Expand
View Pricing
204730
Titanium(IV) oxide, rutile,
99.995% trace metals basis
Expand
View Pricing
635057
Titanium(IV) oxide, rutile,
<001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Expand
View Pricing